Orders LinkContactUsLinkComponent
Bulb pipettes, two marks, class AS
Bulb pipettes, two marks, class AS
Bulb pipettes, two marks, class AS
Supplier:  HIRSCHMANN
Certificates
Bulb pipettes, two marks, class AS
Bulb pipettes, two marks, class AS
11 Options Available Below
Product Details & Documents
AR-Glas®
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in amber stain
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
DIN 12691
Certifications: DIN EN ISO 648; DE-M marking
Product Family Options
Product Information
  • Capacity
    Tolerance
    Length
    Suppl. to DIN
    Availability
    Price
  • 0,5 ml
    0,005 ± ml
    300 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,40 €
    10,40 €
    1 * 1 ST
       10,40 €
  • 1 ml
    0,008 ± ml
    325 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,70 €
    9,70 €
    1 * 1 ST
       9,70 €
  • 2 ml
    0,01 ± ml
    350 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,80 €
    9,80 €
    1 * 1 ST
       9,80 €
  • 3 ml
    0,01 ± ml
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,30 €
    10,30 €
    1 * 1 ST
       10,30 €
  • 5 ml
    0,015 ± ml
    410 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,20 €
    11,20 €
    1 * 1 ST
       11,20 €
  • 10 ml
    0,02 ± ml
    450 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,00 €
    12,00 €
    1 * 1 ST
       12,00 €
  • 15 ml
    0,03 ± ml
    520 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,30 €
    14,30 €
    1 * 1 ST
       14,30 €
  • 20 ml
    0,03 ± ml
    520 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,30 €
    15,30 €
    1 * 1 ST
       15,30 €
  • 25 ml
    0,03 ± ml
    530 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,30 €
    15,30 €
    1 * 1 ST
       15,30 €
  • 50 ml
    0,05 ± ml
    550 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,70 €
    18,70 €
    1 * 1 ST
       18,70 €
  • 100 ml
    0,08 ± ml
    600 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,40 €
    26,40 €
    1 * 1 ST
       26,40 €