ContactUsLinkComponent
Bulb pipettes with syringe, one mark
Bulb pipettes with syringe, one mark
Catalog # 612-1347
Supplier:  HIRSCHMANN
undefined
Bulb pipettes with syringe, one mark
Catalog # 612-1347
Supplier:  HIRSCHMANN
Specifications
  • Capacity:
    50 ml
  • Pk:
    1
  • Tolerance:
    0,05 ± ml
Specifications
About this item
Glass.
  • Silver diffusion ink, durable brown graduations
  • Attached syringe for simple, risk-free pipetting