Accessories for mops micromop™ LoopMop™ and PolyGen™
About this item
Universal electropolished stainless steel handles with a quick disconnect connection that works with the entire range of Micronova adapters, tools and mop heads for simplified and versatile cleaning.
- Universal handles work with all Quick Disconnect tools and mop adapters
- 304 electropolished stainless steel with polyketone components
- Strong, hard-wearing connections for long lasting use
- Available in a range of fixed or extendable lengths from 8 in (20 cm) to 10 ft (3 m)
- Quick Disconnect end attaches directly to QD mops, tools and adapters
- SSA-3 adapter connects QD handles to plastic threaded mops, tools and adapters
- Use the Adjustable Angle Adapters to reach over, under and around obstacles in the cleanroom
- Good compatibility with disinfectants
- Autoclavable
The Micronova Stainless Steel Universal Handles attach directly to the stainless steel Quick Disconnect (QD series) adapters and tools. Use the SSA-3 Quick Disconnect Adapter to attach to the plastic ACME threaded (QP series) adapters and tools. The Adjustable Angle Adapters can be used to create an adjustable angled shape between a handle and mop adapter to clean above and below fixtures, as well as, inside isolators and containment hoods.
2 Options Available Below
Product Details & Documents
Universal electropolished stainless steel handles with a quick disconnect connection that works with the entire range of Micronova adapters, tools and mop heads for simplified and versatile cleaning.
- Universal handles work with all Quick Disconnect tools and mop adapters
- 304 electropolished stainless steel with polyketone components
- Strong, hard-wearing connections for long lasting use
- Available in a range of fixed or extendable lengths from 8 in (20 cm) to 10 ft (3 m)
- Quick Disconnect end attaches directly to QD mops, tools and adapters
- SSA-3 adapter connects QD handles to plastic threaded mops, tools and adapters
- Use the Adjustable Angle Adapters to reach over, under and around obstacles in the cleanroom
- Good compatibility with disinfectants
- Autoclavable
The Micronova Stainless Steel Universal Handles attach directly to the stainless steel Quick Disconnect (QD series) adapters and tools. Use the SSA-3 Quick Disconnect Adapter to attach to the plastic ACME threaded (QP series) adapters and tools. The Adjustable Angle Adapters can be used to create an adjustable angled shape between a handle and mop adapter to clean above and below fixtures, as well as, inside isolators and containment hoods.
Product Family Options
Product Information
- LengthAvailabilityPrice
- 129-0165MicroMop® handle, Fibreglass/PP
- 129-0170MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesMicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesLog in for availability1 * 1 ST137,00 €