Orders LinkContactUsLinkComponent
Mops, PolyGen™
Mops, PolyGen™
   Sustainable Options Available
Supplier:   MICRONOVA
Mops, PolyGen™
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Mops, PolyGen™
2 Options Available Below
Product Details & Documents
The dual-action abrasive mesh is specially adapted to pharmaceutical and biotech projects. Heads used directly with the fibreglass/PP handle or attached to the universal handle via the quick disconnect adapter.
  • Mop can clean to a dry finish or apply disinfectants requiring extended drying time
  • Special herringbone weave ensures no raw edges to guarantee low particulation
  • Low-linting polyester looped strands
  • Reusable, launderable, and autoclavable
Ordering information: Mop heads and handle must be ordered separately.
Specifications for Product Family
Specifications
  • Catalog No:
  • 129-0167
  • 129-0520
  • Environmentally Preferable:
  • Waste Reducing
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 129-0167
    Mop head
    Mop head
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,10 €
    84,10 €
    1 * 1 ST
       84,10 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-0167
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 129-0520
    Mop head, sterile
    Mop head, sterile
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,80 €
    94,80 €
    1 * 1 ST
       94,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    129-0520
    Environmentally Preferable
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    129-0170
    Log in for availability
    1 * 1 ST - 137,00 €
Product Information
  • Catalog #
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    Stainless steel quick disconnect extendable handle
    129-0551
    965 - 1829 mm
    Stainless steel 304
    Log in for availability
    1 * 1 ST - 314,00 €