Orders LinkContactUsLinkComponent
Mops, LoopMop™
Mops, LoopMop™
   Sustainable Options Available
Supplier:   MICRONOVA
Mops, LoopMop™
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Mops, LoopMop™
2 Options Available Below
Product Details & Documents
Knitted polyester mop for applying disinfectant in pharmaceutical, aseptic processing and medical device applications. Absorbent enough to clean to a dry finish but will leave enough moisture for disinfectants that require an extended drying time. Mop heads can be used directly with the fibreglass/polypropylene handle or attached to the universal handle via the quick disconnect adapter.
  • Can be laundered or autoclaved
  • Low linting
  • No raw edges
Ordering information: All components must be ordered separately.
Specifications for Product Family
Specifications
  • Catalog No:
  • 129-0241
  • MICRMZLM5-12IR
  • Environmentally Preferable:
  • Waste Reducing
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 129-0241
    LoopMop™
    LoopMop™
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,80 €
    58,80 €
    1 * 1 ST
       58,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    129-0241
    Environmentally Preferable
  • MICRMZLM5-12IR
    LoopMop™, irradiated
    LoopMop™, irradiated
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 1 620,00 €
    1 620,00 €
    1 * 24 ST
       1 620,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRMZLM5-12IR
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Accessories
Product Information
  • Catalog #
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    Stainless steel quick disconnect extendable handle
    129-0551
    965 - 1829 mm
    Stainless steel 304
    Log in for availability
    1 * 1 ST - 314,00 €
Product Information
  • Catalog #
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect handle
    Stainless steel quick disconnect handle
    129-0062
    155 cm
    Fibreglass
    Log in for availability
    1 * 1 ST - 144,00 €
Product Information
  • Catalog #
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect handle
    Stainless steel quick disconnect handle
    129-0176
    1295 cm
    Stainless steel 304
    Log in for availability
    1 * 1 ST - 140,00 €
Product Information
  • Catalog #
    Availability
    Price
  • MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    129-0170
    Log in for availability
    1 * 1 ST - 137,00 €