Orders LinkContactUsLinkComponent
Slide mailers
Slide mailers
Supplier:   KARTELL
Slide mailers
Supplier:  KARTELL
Certificates
Slide mailers
3 Options Available Below
Product Details & Documents
These mailers are suitable for the safe transportation of cytological and geological microscope slides.
  • Separate compartment with guide rails and more space for each slide
  • Flush closing
  • Lightweight and flat
W×D×H: 50×100×6 mm (for 1); 67×85×6 mm (for 2); 84×100×6 mm (for 3)
Product Family Options
Product Information
  • Colour
    Material
    W×D×H
    No. of slides
    Availability
    Price
  • 631-9160
    Slide mailers
    Slide mailers
    Translucent
    PE
    50×100×6 mm
    1
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 35,50 €
    35,50 €
    1 * 10 ST
       35,50 €
  • 631-9145
    Slide mailers
    Slide mailers
    Translucent
    PP
    67×85×6 mm
    2
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,80 €
    36,80 €
    1 * 10 ST
       36,80 €
  • 631-9161
    Slide mailers
    Slide mailers
    Translucent
    PP
    84×100×6 mm
    3
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 39,70 €
    39,70 €
    1 * 10 ST
       39,70 €