Orders LinkContactUsLinkComponent
VWR®, Slide Folder
VWR®, Slide Folder
Supplier:   VWR Collection
VWR®, Slide Folder
Supplier:  VWR Collection
Certificates
VWR®, Slide Folder
VWR®, Slide Folder
2 Options Available Below
Product Details & Documents
Slide trays hold 20 or 30 standard microscopy slides (76×26 mm).
  • Thumb-cut for easy slide removal
  • Horizontal compartments are recessed, protecting the specimens when the cover is closed
Colour: White (black rims)
Product Family Options
Product Information
  • Colour
    Material
    No. of slides
    Availability
    Price
  • 631-0689
    Slide folder
    Slide folder
    White/black
    Cardboard
    20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,60 €
    11,60 €
    1 * 1 ST
       11,60 €
  • 631-0690
    Slide folder
    Slide folder
    White/black
    Cardboard
    30
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,90 €
    18,90 €
    1 * 1 ST
       18,90 €