Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Supplier:   Epredia
Cavity slides
Supplier:  Epredia
Certificates
Cavity slides
12 Options Available Below
Product Details & Documents
These autoclavable slides are specially designed for immunofluorescence.
  • Adhesive surface and hydrophobic mask
  • Hydrophobic epoxy- or PTFE-coating
  • (*) with rings or wells
Developped for clinical diagnosis applications.
Ordering information: Available with rings or wells (*).
Product Family Options
Epoxy coating
Product Information
  • Colour
    No. of wells
    Well Ø
    Availability
    Price
  • 631-0463
    Cavity slides with epoxy coating
    Cavity slides with epoxy coating
    White
    1
    8 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 129,00 €
    129,00 €
    1 * 100 ST
       129,00 €
  • 631-0464
    Cavity slides with epoxy coating
    Cavity slides with epoxy coating
    White
    1
    15 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 129,00 €
    129,00 €
    1 * 100 ST
       129,00 €
  • 631-0455
    Cavity slides with epoxy coating
    Cavity slides with epoxy coating
    Black
    3
    10 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 129,00 €
    129,00 €
    1 * 100 ST
       129,00 €
  • 631-0453
    Cavity slides with epoxy coating
    Cavity slides with epoxy coating
    Black
    3
    14 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 129,00 €
    129,00 €
    1 * 100 ST
       129,00 €
  • 631-0451
    Cavity slides with epoxy coating
    Cavity slides with epoxy coating
    Black
    8
    6 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 129,00 €
    129,00 €
    1 * 100 ST
       129,00 €
  • 631-0457
    Cavity slides with epoxy coating
    Cavity slides with epoxy coating
    Black
    10
    6 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 129,00 €
    129,00 €
    1 * 100 ST
       129,00 €
  • 631-0452
    Cavity slides with epoxy coating
    Cavity slides with epoxy coating
    White
    12
    5 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 129,00 €
    129,00 €
    1 * 100 ST
       129,00 €
  • 631-0460
    Cavity slides with epoxy coating
    Cavity slides with epoxy coating
    Black
    21
    4 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 129,00 €
    129,00 €
    1 * 100 ST
       129,00 €
PTFE coating
Product Information
  • Colour
    No. of wells
    Well Ø
    Availability
    Price
  • 631-0467
    Cavity slides with PTFE coating
    Cavity slides with PTFE coating
    Black
    3
    14 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 191,00 €
    191,00 €
    1 * 100 ST
       191,00 €
  • 631-0465
    Cavity slides with PTFE coating
    Cavity slides with PTFE coating
    Black
    8
    6 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 191,00 €
    191,00 €
    1 * 100 ST
       191,00 €
  • 631-1371
    Cavity slides with PTFE coating
    Cavity slides with PTFE coating
    Black
    10
    6 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 191,00 €
    191,00 €
    1 * 100 ST
       191,00 €
  • 631-9423
    Cavity slides with PTFE coating
    Cavity slides with PTFE coating
    White
    12
    5 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 191,00 €
    191,00 €
    1 * 100 ST
       191,00 €