Orders LinkContactUsLinkComponent
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
Supplier:  VWR Collection
Certificates
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
13 Options Available Below
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: In plastic hinged lid boxes.
Product Family Options
Product Information
  • L×W
    No.
    Availability
    Price
  • 18×24 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 39,30 €
    39,30 €
    1 * 1.000 ST
       39,30 €
  • 20×20 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 30,60 €
    30,60 €
    1 * 1.000 ST
       30,60 €
  • 22×32 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 54,20 €
    54,20 €
    1 * 1.000 ST
       54,20 €
  • 22×32 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 61,30 €
    61,30 €
    1 * 1.000 ST
       61,30 €
  • 22×40 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 67,00 €
    67,00 €
    1 * 1.000 ST
       67,00 €
  • 22×40 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 75,70 €
    75,70 €
    1 * 1.000 ST
       75,70 €
  • 22×50 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 77,70 €
    77,70 €
    1 * 1.000 ST
       77,70 €
  • 22×50 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 87,90 €
    87,90 €
    1 * 1.000 ST
       87,90 €
  • 24×32 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 55,20 €
    55,20 €
    1 * 1.000 ST
       55,20 €
  • 24×32 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 62,80 €
    62,80 €
    1 * 1.000 ST
       62,80 €
  • 24×40 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 67,20 €
    67,20 €
    1 * 1.000 ST
       67,20 €
  • 24×50 mm
    #1
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,10 €
    73,10 €
    1 * 1.000 ST
       73,10 €
  • 24×50 mm
    #1,5
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 89,60 €
    89,60 €
    1 * 1.000 ST
       89,60 €