Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
Cover glasses
Supplier:  MARIENFELD
Certificates
Cover glasses
16 Options Available Below
Product Details & Documents
Made of chemically resistant borosilicate D 263™ M glass, Hydrolytic Class I
  • No bubbles or flaws
  • Suitable for in vitro diagnostic (IVD) applications
Thickness: 0,13 - 0,16 mm
Packaging: Supplied 100 in hinged boxes or in boxes with separate lid.
Documents
Product Family Options
Plastic boxes with separate lid
Product Information
  • L×W
    No.
    Material
    Availability
    Price
  • 8×8 mm
    #2
    Glass
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • 12×12 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 26,50 €
    26,50 €
    1 * 1.000 ST
       26,50 €
  • 15×15 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 27,00 €
    27,00 €
    1 * 1.000 ST
       27,00 €
  • 16×16 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 26,80 €
    26,80 €
    1 * 1.000 ST
       26,80 €
  • 18×18 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 23,60 €
    23,60 €
    1 * 1.000 ST
       23,60 €
  • 20×20 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 31,10 €
    31,10 €
    1 * 1.000 ST
       31,10 €
  • 22×22 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 32,40 €
    32,40 €
    1 * 1.000 ST
       32,40 €
  • 24×24 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 41,10 €
    41,10 €
    1 * 1.000 ST
       41,10 €
Plastic boxes with hinged lid
Product Information
  • L×W
    No.
    Material
    Availability
    Price
  • 21×26 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 96,60 €
    96,60 €
    1 * 2.000 ST
       96,60 €
  • 22×40 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 62,60 €
    62,60 €
    1 * 1.000 ST
       62,60 €
  • 22×50 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 72,80 €
    72,80 €
    1 * 1.000 ST
       72,80 €
  • 22×60 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 81,20 €
    81,20 €
    1 * 1.000 ST
       81,20 €
  • 24×24 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 83,20 €
    83,20 €
    1 * 2.000 ST
       83,20 €
  • 24×36 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 61,90 €
    61,90 €
    1 * 1.000 ST
       61,90 €
  • 24×46 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 73,30 €
    73,30 €
    1 * 1.000 ST
       73,30 €
  • 24×48 mm
    #1
    Glass
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 75,40 €
    75,40 €
    1 * 1.000 ST
       75,40 €