Labels for slides
These thin labels are suitable for use with microscopy slides.
- Permanent adhesive
- No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
These thin labels are suitable for use with microscopy slides.
- Permanent adhesive
- No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
Product Family Options
- DescriptionMaterialColourDimensionsPacked
- Slide labels on sheets, custom imprintPaperBlack on white11×22 mm1 * 1.000 ST119,00 €
- Slide labels on sheetsPaperBlack on white22×22 mm1 * 1.000 ST35,00 €
- Slide labels on sheets, custom imprintPaperBlack on white22×22 mm1 * 1.000 ST125,00 €
- Slide labels on sheets, custom imprint, tagPaperBlack on white22×22 mm1 * 1.000 ST35,50 €
- Slide labelsPaperBlack on white22×22 mmOn roll1 * 1.000 ST35,00 €
- Slide labels on roll, custom imprintPaperBlack on white22×22 mmOn roll1 * 1.000 ST125,00 €