Orders LinkContactUsLinkComponent
Microscope slides, UniMark®
Microscope slides, UniMark®
Microscope slides, UniMark®
Supplier:  MARIENFELD
Certificates
Microscope slides, UniMark®
Microscope slides, UniMark®
12 Options Available Below
Product Details & Documents
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
  • Frosted end for labelling
  • Soda-lime glass, Hydrolytic Class 3
  • Different colours to identify preparations securely
  • Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
Documents
Certificates
Search by Lot Number
Product Family Options
Ground edges
Product Information
  • Colour
    Availability
    Price
  • 631-0093
    Slides in standard packs
    Slides in standard packs
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 7,50 €
    7,50 €
    1 * 50 ST
       7,50 €
  • 631-0094
    Slides in standard packs
    Slides in standard packs
    Yellow
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 7,50 €
    7,50 €
    1 * 50 ST
       7,50 €
  • 631-0095
    Slides in standard packs
    Slides in standard packs
    Green
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 7,50 €
    7,50 €
    1 * 50 ST
       7,50 €
  • 631-0096
    Slides in standard packs
    Slides in standard packs
    Blue
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 7,50 €
    7,50 €
    1 * 50 ST
       7,50 €
  • 631-0097
    Slides in standard packs
    Slides in standard packs
    Rose
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 7,50 €
    7,50 €
    1 * 50 ST
       7,50 €
  • 631-1209
    Slides in standard packs
    Slides in standard packs
    Orange
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 7,50 €
    7,50 €
    1 * 50 ST
       7,50 €
Cut edges
Product Information
  • Colour
    Availability
    Price
  • 631-1203
    Slides in standard packs
    Slides in standard packs
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 5,20 €
    5,20 €
    1 * 50 ST
       5,20 €
  • 631-1204
    Slides in standard packs
    Slides in standard packs
    Yellow
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 5,20 €
    5,20 €
    1 * 50 ST
       5,20 €
  • 631-1205
    Slides in standard packs
    Slides in standard packs
    Green
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 5,20 €
    5,20 €
    1 * 50 ST
       5,20 €
  • 631-1206
    Slides in standard packs
    Slides in standard packs
    Blue
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 5,20 €
    5,20 €
    1 * 50 ST
       5,20 €
  • 631-1207
    Slides in standard packs
    Slides in standard packs
    Rose
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 5,20 €
    5,20 €
    1 * 50 ST
       5,20 €
  • 631-1208
    Slides in standard packs
    Slides in standard packs
    Orange
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 5,20 €
    5,20 €
    1 * 50 ST
       5,20 €