ContactUsLinkComponent
PCR plates, 384-well, Thermo-Fast® 384 Mark II
PCR plates, 384-well, Thermo-Fast® 384 Mark II
Catalog # 732-4912
undefined
PCR plates, 384-well, Thermo-Fast® 384 Mark II
Catalog # 732-4912
Specifications
  • No. of wells:
    384
  • Profile:
    Standard
  • Skirt:
    Fully skirted
  • Well colour:
    Clear
  • Pk:
    100
  • Well volume:
    0,04 ml
Specifications
About this item
Moulded from ultra low retention PP that assists in preventing adhesion of samples to the side walls. Due to the increased well volume, post-PCR applications such as enzymatic digestions and ethanol precipitation can be carried out in the wells. Compatible with all leading 384 block thermal cyclers.
  • Fully skirted for compatibility with automated systems
  • 8 holes in the skirt aid plate positioning and removal from the thermal cycler block
  • Cut away corner aids orientation
  • Certified free from DNase, RNase and human genomic DNA
Maximum well volume: 55 µl
Working volume: 40 µl