polypropylene storage plates 96 well, Falcon®
Supplier: Corning
Certificates
About this item
PP plate, PP lid, non sterile. Plates designed for compound storage.
- Rigid and flat plate for consistent automated handling
- Low retention PP and well design give low residual volumes after pipetting
- Lid compatible with automated lidding and de-lidding devices
- Raised well edges and flatness for heat and adhesive sealing
Frequently Bought Together