CovaLink™ NH modules, Nunc™
Catalog # 735-0153
Fournisseur: Thermo Fisher Scientific
Spécifications
- Colour:Clear
- Description:F8×12 module, clear, framed
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Pk:30
Spécifications
À propos de cet article
PS. Developed to bind compounds covalently to plastic surfaces. For use with molecules which do not adsorb easily to a traditional surface. Framed modules with alphanumeric orientation (external dimensions: 128×86 mm) fit all standard equipment.
- Very thorough washing possible with high ionic strength solutions
- Minimum non-specific binding
- Allows site-specific binding via, for example, a carboxylic group of peptide or phosphate group in the 5' end of nucleic acids
- Free-standing modules with flat (F) wells
Recommended working volume: 100 µl
Recommended working volume: 100 µl