Orders LinkContactUsLinkComponent
Microarray plates, HTA™ Plate
Microarray plates, HTA™ Plate
Supplier:   Greiner Bio-One
Microarray plates, HTA™ Plate
Supplier:  Greiner Bio-One
Certificates
Microarray plates, HTA™ Plate
Microarray plates, HTA™ Plate
3 Options Available Below
Product Details & Documents
Plastic plates for high-throughput microarrays. With 96 shallow wells and a low well rim, the HTA™ Plate has been optimised for the simultaneous analysis of multiple samples.
  • Available with multifunctional, 3-D Amino, 3-D Aldehyde or 3-D Epoxy surface functionality
  • Partition of the plate into four individual strips containing 24 wells each gives flexibility, permitting processing of a variable number of samples each day
  • Plate dimensions are compatible with automated systems
  • Low well rim of only 0,3 mm
  • Low autofluorescence
Product Family Options
Product Information
  • Availability
    Price
  • 731-3018
    HTA Plate, 3-D Amino
    HTA Plate, 3-D Amino
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 1 670,00 €
    1 670,00 €
    1 * 5 ST
       1 670,00 €
  • 731-3019
    HTA Plate, 3-D Aldehyde
    HTA Plate, 3-D Aldehyde
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 1 670,00 €
    1 670,00 €
    1 * 5 ST
       1 670,00 €
  • 731-3020
    HTA Plate, 3-D Epoxy
    HTA Plate, 3-D Epoxy
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 1 670,00 €
    1 670,00 €
    1 * 5 ST
       1 670,00 €