Orders LinkContactUsLinkComponent
96-Multiwell insert system, Falcon®, Corning
96-Multiwell insert system, Falcon®, Corning
   Sustainable Options Available
Supplier:   Corning
96-Multiwell insert system, Falcon®, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
96-Multiwell insert system, Falcon®, Corning
2 Options Available Below
Product Details & Documents
The Falcon® HTS 96-multiwell insert system is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies. This automation compatible platform is composed of a 1,0 µm pore size PET membrane-based 96-multiwell insert plate, a media feeder tray and a lid. The drop-in baffle for the feeder tray mitigates media sloshing and lowers the risk of contamination.
  • Automation compatible design suitable for use with most robots and fluid handling instruments
  • Excellent reproducibility - the one-piece feeder tray enhances consistency in well-to-well monolayer growth
  • Ideal for drug transport studies
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-0368
  • 734-1379
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 - Bedford
  • ISO 14001 - Bedford
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 734-0368
    Five insert plates with feeder trays and lids
    Five insert plates with feeder trays and lids
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 2 582,07 €
    2 582,07 €
    1 * 5 ST
       2 582,07 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-0368
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 - Bedford
      Low Manufacturing Impact
  • 734-1379
    Five insert plates with 96-square well, angled-bottom plates and lids
    Five insert plates with 96-square well, angled-bottom plates and lids
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 2 740,30 €
    2 740,30 €
    1 * 5 ST
       2 740,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1379
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 - Bedford
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • 96-square well, angled-bottom plate, with lid
    96-square well, angled-bottom plate, with lid
    734-1378
    Log in for availability
    1 * 5 ST - 531,09 €
Product Information
  • Catalog #
    Availability
    Price
  • Feeder tray, PS, non-treated, designed for use with Falcon® 96-multiwell insert systems
    Feeder tray, PS, non-treated, designed for use with Falcon® 96-multiwell insert systems
    736-2024
    Log in for availability
    1 * 5 ST - 187,12 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.