ContactUsLinkComponent
Filter plates, 384-well, 100 µl, AcroPrep™
Filter plates, 384-well, 100 µl, AcroPrep™
Certificates
undefined
Filter plates, 384-well, 100 µl, AcroPrep™
Specifications
  • Catalog No:
  • 516-0173
  • 516-0174
  • 516-0175
  • 516-0207
  • 736-4006
  • 516-0170
  • 736-4005
  • 516-0176
  • 736-4004
  • 516-0172
  • Model:
  • AcroPrep™ 384, 100 μl (short tips)
  • AcroPrep™ 384, 100 μl (long tips)
  • AcroPrep™ 384, 100 μl (short tips)
  • AcroPrep™ 384, 100 μl (short tips)
  • AcroPrep™ 384, 100 μl (short tips)
  • AcroPrep™ 384, 100 μl (long tips)
  • AcroPrep™ 384, 100 μl (short tips)
  • AcroPrep™ 384, 100 μl (long tips)
  • AcroPrep™ 384, 100 μl (long tips)
  • AcroPrep™ 384, 100 μl (long tips)
  • Typical hold-up volume:
  • < 4 µl
  • Well bottom area:
  • 0,05 cm²
  • 0,05 cm²
  • 0,05 cm²
  • 0,05 cm²
  • 0,05 cm²
  • 0,05 cm²
  • 0,05 cm²
  • 0,05 cm²
  • 0,05 cm²
  • 0,05 cm²
  • Max. working volume/well:
  • 80 µl
  • 80 µl
  • 80 µl
  • 80 µl
  • 80 µl
  • 80 µl
  • 80 µl
  • 80 µl
  • 80 µl
  • 80 µl
  • L×W×H with lid:
  • 128×86×17 mm
  • 128×86×21 mm
  • 128×86×17 mm
  • 128×86×17 mm
  • 128×86×17 mm
  • 128×86×21 mm
  • 128×86×17 mm
  • 128×86×21 mm
  • 128×86×21 mm
  • 128×86×21 mm
  • L×W×H without lid:
  • 128×86×14 mm
  • 128×86×19 mm
  • 128×86×14 mm
  • 128×86×14 mm
  • 128×86×14 mm
  • 128×86×19 mm
  • 128×86×14 mm
  • 128×86×19 mm
  • 128×86×19 mm
  • 128×86×19 mm