ContactUsLinkComponent
Invasion chambers, Corning® BioCoat™ Matrigel®
Invasion chambers, Corning® BioCoat™ Matrigel®
Supplier:  Corning
   Sustainable Options Available
Certificates
undefined
Invasion chambers, Corning® BioCoat™ Matrigel®
Supplier:  Corning
   Sustainable Options Available
Certificates
Specifications
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Quality control:
    Tested for the ability to allow invasion of HT-1080 cells, an invasive human fibrosarcoma cell line, and to exclude invasion of 3T3 cells, a non-invasive mouse fibroblast cell line
    Tested and found negative for bacteria and fungi
  • Storage and stability:
    Store at −20 °C in original packaging. Stable for at least 3 months at −20 °C