Orders LinkContactUsLinkComponent
Microplates, 384-well
Microplates, 384-well
384-well plate, Clear, 384, -
Catalog # 735-0577
   Low Manufacturing Impact
Microplates, 384-well
Catalog # 735-0577
   Low Manufacturing Impact
Microplates, 384-well
Microplates, 384-well
384-well plate, Clear, 384, -
Price
359,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Ideal for high throughput screening, these 384-well microplates allow miniaturisation of assays and many other applications, including high volume DNA library manipulation and solid phase binding assays. Available with a choice of clear microplates for colorimetric applications, white plates for luminometric/fluorometric applications, and black microplates for fluorometric applications. There are two different designs, both of which incorporate flat bottom wells and a standard microplate footprint. The first type has a square well shape with rounded corners for an increased well volume. It has the standard microplate height and a total well volume of 120 µl. The second type has round wells and a low profile, with a total well volume of 70 µl.
  • Flat bottom wells for optimum optical characteristics
  • Standard footprint compatible with automated handling equipment
  • Choice of rounded square or round well shape
Specifications
  • No. of wells:
    384
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    384-well plate
  • Cat. No.:
    735-0577
  • Environmental certification:
    ISO 14001 (Certificate Registration No. EMS 623400)
  • Environmentally Preferable:
    Low Manufacturing Impact