Orders LinkContactUsLinkComponent
Microplates, 96-well
   Sustainable Options Available
Supplier:   Greiner Bio-One
Microplates, 96-well
Supplier:  Greiner Bio-One
   Sustainable Options Available
Certificates
Microplates, 96-well
16 Options Available Below
Product Details & Documents
PP, untreated, without lid. These microplates are ideal for storage of DNA or RNA, stock cultures, and long-term storage of active agents. Versions available sterile or non sterile and with round, conical or flat bottom well shape.
  • Uniform external dimensions with 9 mm well to well spacing
  • Alphanumeric well coding
  • High chemical and temperature resistance (−196 to +121 °C)
  • Sealable with adhesive films, heat seals or CapMats
  • RNase- and DNase-free
Specifications for Product Family
Specifications
  • Catalog No:
  • 736-0116
  • 736-0117
  • 738-0006
  • 738-0007
  • GREI650203_100
  • GREI650204_100
  • GREI650205_100
  • GREI650206_100
  • 736-0118
  • 738-0011
  • GREI651261_100
  • GREI651203_100
  • GREI651204_100
  • 736-0121
  • 736-0122
  • 738-0026
  • Model:
  • 736-0116
  • 736-0117
  • 738-0006
  • 738-0007
  • 736-0118
  • 738-0011
  • 736-0121
  • 736-0122
  • 738-0026
  • Total volume:
  • 355 µl
  • 355 µl
  • 355 µl
  • 355 µl
  • 355 µl
  • 355 µl
  • 355 µl
  • 355 µl
  • 340 µl
  • 340 µl
  • 340 µl
  • 340 µl
  • 340 µl
  • 392 µl
  • 392 µl
  • 392 µl
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Well profile:
  • U-bottom
  • U-bottom
  • U-bottom
  • U-bottom
  • U-bottom
  • U-bottom
  • U-bottom
  • U-bottom
  • V-bottom
  • V-bottom
  • V-bottom
  • V-bottom
  • V-bottom
  • Flat bottom
  • Flat bottom
  • Flat bottom
  • Working volume:
  • 50 - 300 µl
  • 50 - 300 µl
  • 50 - 300 µl
  • 50 - 300 µl
  • 50 - 300 µl
  • 50 - 300 µl
  • 50 - 300 µl
  • 50 - 300 µl
  • 50 - 335 µl
  • 50 - 335 µl
  • 50 - 335 µl
  • 50 - 335 µl
  • 50 - 335 µl
  • 25 - 370 µl
  • 25 - 370 µl
  • 25 - 370 µl
  • Environmental certification:
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
Product Information
  • Colour
    No. of wells
    Sterile
    Availability
    Price
  • 736-0116
    U-bottom (chimney well), natural
    U-bottom (chimney well), natural
    Natural
    96
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 210,00 €
    210,00 €
    1 * 100 ST
       210,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    736-0116
    Model
    736-0116
    Total volume
    355 µl
    Environmentally Preferable
    Well profile
    U-bottom
    Working volume
    50 - 300 µl
    Environmental certification
  • 736-0117
    U-bottom (chimney well), natural
    U-bottom (chimney well), natural
    Natural
    96
    +
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 272,00 €
    272,00 €
    1 * 100 ST
       272,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    736-0117
    Model
    736-0117
    Total volume
    355 µl
    Environmentally Preferable
    Well profile
    U-bottom
    Working volume
    50 - 300 µl
    Environmental certification
  • 738-0006
    U-bottom (chimney well), natural
    U-bottom (chimney well), natural
    White
    96
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 318,00 €
    318,00 €
    1 * 100 ST
       318,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    738-0006
    Model
    738-0006
    Total volume
    355 µl
    Environmentally Preferable
    Well profile
    U-bottom
    Working volume
    50 - 300 µl
    Environmental certification
  • 738-0007
    U-bottom (chimney well), natural
    U-bottom (chimney well), natural
    Black
    96
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 296,00 €
    296,00 €
    1 * 100 ST
       296,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    738-0007
    Model
    738-0007
    Total volume
    355 µl
    Environmentally Preferable
    Well profile
    U-bottom
    Working volume
    50 - 300 µl
    Environmental certification
  • GREI650203_100
    U-bottom (chimney well), natural
    U-bottom (chimney well), natural
    Red
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 300,00 €
    300,00 €
    1 * 100 ST
       300,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    GREI650203_100
    Model
    Total volume
    355 µl
    Environmentally Preferable
    Well profile
    U-bottom
    Working volume
    50 - 300 µl
    Environmental certification
  • GREI650204_100
    U-bottom (chimney well), natural
    U-bottom (chimney well), natural
    Blue
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 285,00 €
    285,00 €
    1 * 100 ST
       285,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    GREI650204_100
    Model
    Total volume
    355 µl
    Environmentally Preferable
    Well profile
    U-bottom
    Working volume
    50 - 300 µl
    Environmental certification
  • GREI650205_100
    U-bottom (chimney well), natural
    U-bottom (chimney well), natural
    Green
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 300,00 €
    300,00 €
    1 * 100 ST
       300,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    GREI650205_100
    Model
    Total volume
    355 µl
    Environmentally Preferable
    Well profile
    U-bottom
    Working volume
    50 - 300 µl
    Environmental certification
  • GREI650206_100
    U-bottom (chimney well), natural
    U-bottom (chimney well), natural
    Yellow
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 300,00 €
    300,00 €
    1 * 100 ST
       300,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    GREI650206_100
    Model
    Total volume
    355 µl
    Environmentally Preferable
    Well profile
    U-bottom
    Working volume
    50 - 300 µl
    Environmental certification
  • 736-0118
    V-bottom (chimney well)
    V-bottom (chimney well)
    Natural
    96
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 195,00 €
    195,00 €
    1 * 100 ST
       195,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0118
    Model
    736-0118
    Total volume
    340 µl
    Environmentally Preferable
    Low Manufacturing Impact
    Well profile
    V-bottom
    Working volume
    50 - 335 µl
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0011
    V-bottom (chimney well)
    V-bottom (chimney well)
    Black
    96
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 291,00 €
    291,00 €
    1 * 100 ST
       291,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0011
    Model
    738-0011
    Total volume
    340 µl
    Environmentally Preferable
    Low Manufacturing Impact
    Well profile
    V-bottom
    Working volume
    50 - 335 µl
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • GREI651261_100
    V-bottom (chimney well)
    V-bottom (chimney well)
    Natural
    +
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 288,00 €
    288,00 €
    1 * 100 ST
       288,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI651261_100
    Model
    Total volume
    340 µl
    Environmentally Preferable
    Low Manufacturing Impact
    Well profile
    V-bottom
    Working volume
    50 - 335 µl
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • GREI651203_100
    V-bottom (chimney well)
    V-bottom (chimney well)
    Red
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 300,00 €
    300,00 €
    1 * 100 ST
       300,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI651203_100
    Model
    Total volume
    340 µl
    Environmentally Preferable
    Low Manufacturing Impact
    Well profile
    V-bottom
    Working volume
    50 - 335 µl
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • GREI651204_100
    V-bottom (chimney well)
    V-bottom (chimney well)
    Blue
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 300,00 €
    300,00 €
    1 * 100 ST
       300,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI651204_100
    Model
    Total volume
    340 µl
    Environmentally Preferable
    Low Manufacturing Impact
    Well profile
    V-bottom
    Working volume
    50 - 335 µl
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 736-0121
    F-bottom (chimney well)
    F-bottom (chimney well)
    Natural
    96
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 213,00 €
    213,00 €
    1 * 100 ST
       213,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    736-0121
    Model
    736-0121
    Total volume
    392 µl
    Environmentally Preferable
    Well profile
    Flat bottom
    Working volume
    25 - 370 µl
    Environmental certification
  • 736-0122
    F-bottom (chimney well)
    F-bottom (chimney well)
    Natural
    96
    +
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 254,00 €
    254,00 €
    1 * 100 ST
       254,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    736-0122
    Model
    736-0122
    Total volume
    392 µl
    Environmentally Preferable
    Well profile
    Flat bottom
    Working volume
    25 - 370 µl
    Environmental certification
  • 738-0026
    F-bottom (chimney well)
    F-bottom (chimney well)
    Black
    96
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 289,00 €
    289,00 €
    1 * 100 ST
       289,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    738-0026
    Model
    738-0026
    Total volume
    392 µl
    Environmentally Preferable
    Well profile
    Flat bottom
    Working volume
    25 - 370 µl
    Environmental certification