Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
   Sustainable Options Available
Supplier:   Greiner Bio-One
Microplates, 96-well
Supplier:  Greiner Bio-One
   Sustainable Options Available
Certificates
Microplates, 96-well
16 Options Available Below
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specifications for Product Family
Specifications
  • Catalog No:
  • 391-3610
  • 736-0223
  • 391-3605
  • 391-3311
  • 391-3615
  • 736-0119
  • 738-0168
  • 738-0208
  • 736-0228
  • 738-0016
  • 736-0229
  • 391-3629
  • 738-0020
  • 738-0021
  • 738-0022
  • 738-0023
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
96-well microplates with solid bottom
Product Information
  • Recommended working volume
    No. of wells
    Sterile
    Total volume
    Availability
    Price
  • V-bottom, clear
    40 - 200 µl
    96
    -
    234 µl
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 62,10 €
    62,10 €
    1 * 100 ST
       62,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-3610
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 736-0223
    V-bottom, clear, sterile
    V-bottom, clear, sterile
    40 - 200 µl
    96
    +
    234 µl
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 84,60 €
    84,60 €
    1 * 100 ST
       84,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0223
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • U-bottom, clear
    40 - 280 µl
    96
    -
    323 µl
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,70 €
    61,70 €
    1 * 100 ST
       61,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    391-3605
    Environmentally Preferable
    Environmental certification
  • 391-3311
    U-bottom, clear, sterile
    U-bottom, clear, sterile
    40 - 280 µl
    96
    +
    323 µl
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 96,10 €
    96,10 €
    1 * 100 ST
       96,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    391-3311
    Environmentally Preferable
    Environmental certification
  • F-bottom, clear
    40 - 340 µl
    96
    -
    382 µl
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,70 €
    61,70 €
    1 * 100 ST
       61,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-3615
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 736-0119
    F-bottom, clear, sterile
    F-bottom, clear, sterile
    40 - 340 µl
    96
    +
    382 µl
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 90,30 €
    90,30 €
    1 * 100 ST
       90,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0119
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0168
    F-bottom, clear
    F-bottom, clear
    40 - 340 µl
    -
    382 µl
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 26,40 €
    26,40 €
    1 * 50 ST
       26,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0168
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0208
    V-bottom, clear, non sterile
    V-bottom, clear, non sterile
    40 - 200 µl
    -
    234 µl
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 306,00 €
    306,00 €
    1 * 400 ST
       306,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0208
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
96-well microplates with solid bottom, F-bottom (chimney well) well profile
Product Information
  • Recommended working volume
    No. of wells
    Sterile
    Total volume
    Availability
    Price
  • 736-0228
    White, LUMITRAC™ 200 medium binding
    White, LUMITRAC™ 200 medium binding
    40 - 340 µl
    96
    -
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 106,00 €
    106,00 €
    1 * 40 ST
       106,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0228
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0016
    White, LUMITRAC™ 600 high binding, sterile
    White, LUMITRAC™ 600 high binding, sterile
    40 - 340 µl
    96
    +
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 114,00 €
    114,00 €
    1 * 40 ST
       114,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0016
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 736-0229
    Black, FLUOTRAC™ 200 medium binding
    Black, FLUOTRAC™ 200 medium binding
    40 - 340 µl
    96
    -
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 153,00 €
    153,00 €
    1 * 40 ST
       153,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    736-0229
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 391-3629
    Black, FLUOTRAC™ 600 high binding, sterile
    Black, FLUOTRAC™ 600 high binding, sterile
    40 - 340 µl
    96
    +
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 115,00 €
    115,00 €
    1 * 40 ST
       115,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-3629
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
96-well microplates with µClear® bottom, F-bottom (chimney well) well profile
Product Information
  • Recommended working volume
    No. of wells
    Sterile
    Total volume
    Availability
    Price
  • 738-0020
    White, high binding, sterile
    White, high binding, sterile
    40 - 340 µl
    96
    +
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 242,00 €
    242,00 €
    1 * 40 ST
       242,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0020
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0021
    White, medium binding
    White, medium binding
    40 - 340 µl
    96
    -
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 243,00 €
    243,00 €
    1 * 40 ST
       243,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0021
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0022
    Black, medium binding
    Black, medium binding
    40 - 340 µl
    96
    -
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 241,00 €
    241,00 €
    1 * 40 ST
       241,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0022
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0023
    Black, high binding, sterile
    Black, high binding, sterile
    40 - 340 µl
    96
    +
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 241,00 €
    241,00 €
    1 * 40 ST
       241,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0023
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact