Microplates, 96-well
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Documents
- Catalog No:
- 738-0032
- 738-0033
- 736-0227
- GREI655001-000
- 738-0015
- 738-0017
- 391-3390
- 738-0167
- 738-0004
- 738-0005
- 738-0008
- 738-0009
- Total volume:
- 199 µl
- 199 µl
- 382 µl
- 382 µl
- 382 µl
- 392 µl
- 392 µl
- 382 µl
- 323 µl
- 323 µl
- 234 µl
- 234 µl
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Product Family Options
- DescriptionWorking volumeTotal volume
- 96-well microplate, half area, MICROLON® 20015 - 175 µl199 µl1 * 40 ST124,00 €
Low Manufacturing Impact Specifications
Cat. No.738-0032Total volume199 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, half area, MICROLON® 60015 - 175 µl199 µl1 * 40 ST141,00 €
Low Manufacturing Impact Specifications
Cat. No.738-0033Total volume199 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, F-bottom (standard), MICROLON® 20025 - 340 µl382 µl1 * 40 ST64,20 €
Low Manufacturing Impact Specifications
Cat. No.736-0227Total volume382 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, F-bottom (standard), MICROLON® 200, individually packed25 - 340 µl382 µl1 * 40 ST96,40 €
Low Manufacturing Impact Specifications
Cat. No.GREI655001-000Total volume382 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, F-bottom (standard), MICROLON® 60025 - 340 µl382 µl1 * 40 ST69,20 €
Low Manufacturing Impact Specifications
Cat. No.738-0015Total volume382 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, F-bottom (chimney well), MICROLON® 20025 - 340 µl392 µl1 * 40 ST97,70 €
Low Manufacturing Impact Specifications
Cat. No.738-0017Total volume392 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, F-bottom (chimney well), MICROLON® 60025 - 340 µl392 µl1 * 40 ST110,00 €
Low Manufacturing Impact Specifications
Cat. No.391-3390Total volume392 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, F-bottom, MICROLON® 600, low RAF, 2/bag25 - 340 µl382 µl1 * 50 ST136,00 €
Low Manufacturing Impact Specifications
Cat. No.738-0167Total volume382 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, U-bottom, MICROLON® 20040 - 280 µl323 µl1 * 40 ST98,40 €
Specifications
Cat. No.738-0004Total volume323 µlEnvironmentally PreferableEnvironmental certification - 96-well microplate, U-bottom, MICROLON® 60040 - 280 µl323 µl1 * 40 ST110,00 €
Specifications
Cat. No.738-0005Total volume323 µlEnvironmentally PreferableEnvironmental certification - 96-well microplate, V-bottom, MICROLON® 20040 - 200 µl234 µl1 * 40 ST98,40 €
Low Manufacturing Impact Specifications
Cat. No.738-0008Total volume234 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 96-well microplate, V-bottom, MICROLON® 60040 - 200 µl234 µl1 * 40 ST117,00 €
Low Manufacturing Impact Specifications
Cat. No.738-0009Total volume234 µlEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact