ContactUsLinkComponent
Terasaki plates, Nunc™ MiniTray
Terasaki plates, Nunc™ MiniTray
   Sustainable Options Available
Certificates
undefined
Terasaki plates, Nunc™ MiniTray
   Sustainable Options Available
Specifications
  • Catalog No:
  • 735-0201
  • 735-0203
  • 735-0202
  • Model:
  • MiniTray, low profile, with lid
  • MiniTray, high profile, S-plate
  • MiniTray, low profile, with lid
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Total volume:
  • 10 µl
  • 10 µl
  • 10 µl
  • Dimensions:
  • Top: 3,82 mm; Bottom: 1,34 mm; Depth: 1,92 mm
  • Top: 3,72 mm; Bottom: 1,30 mm; Depth: 2,10 mm
  • Top: 3,82 mm; Bottom: 1,34 mm; Depth: 1,92 mm
  • Tray W×L:
  • 81×56 mm (without lid); 84×94 mm (with lid)
  • 81×56 mm (without lid)
  • 81×56 mm (without lid); 84×94 mm (with lid)
  • Environmental certification:
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)