MicroWell™ Plates, 96 well, FluoroNunc™ and LumiNunc™
PS, untreated, pinchbar design. 96-MicroWell™ plates for fluorescence and luminescence based assays. Black plates for fluorescence reading with minimum back-scattered light and background fluorescence. White plates for luminescence with maximum reflection and minimal autoluminescence.
- Raised well rims reduce risk of cross-contamination
- Flat (F) bottom wells for optimum optical characteristics
- Plates fit all standard equipment
- Compatible with lids for MicroWell™ plates and adhesive sealing tape
Recommended working volume: 200 µl per well
Recommended working volume: 200 µl per well
Frequently Bought Together