Immunology plates and modules, Nunc Immobilizer™
Supplier: Thermo Fisher Scientific
Certificates
About this item
PS, non sterile, without lid. Available with a choice of surface chemistries: Amino surface for rapid coupling of peptides and proteins, streptavidin surface for rapid coupling of biotinylated biomolecules such as peptides, antibodies, oligonucleotides or haptens. Also available clear, for colorimetric assays and sample storage, white, for luminescence and fluorescence assays, or black, for fluorescence assays.
- Ready to use plates and modules requiring no activation or blocking steps
- Robust and stable surfaces with long shelf life
- High sensitivity and specificity
- High specific binding
- High signal to noise ratio
Total well volume: 400 µl (96-well plates and flat well strips), 350 µl (C-well strips), 120 µl (384-well plates).
Total well volume: 400 µl (96-well plates and flat well strips), 350 µl (C-well strips), 120 µl (384-well plates)
Specifications
- Catalog No:
- 734-1286
- 734-1285
- 734-1287
- 734-1284
- 734-1291
- 734-1290
- 734-1292
- 734-1279
- 734-1278
- 734-1283
- 734-1277
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)