Orders LinkContactUsLinkComponent
Measuring cylinders, low form, PMP
Measuring cylinders, low form, PMP
Supplier:   KARTELL
Measuring cylinders, low form, PMP
Supplier:  KARTELL
Certificates
Measuring cylinders, low form, PMP
Measuring cylinders, low form, PMP
8 Options Available Below
Product Details & Documents
PMP (TPX®), transparent, low form, autoclavable.
  • Pouring spout, circular base and moulded white graduations
  • Excellent chemical resistance
  • Can withstand temperatures up to +170 °C
Documents
Product Family Options
Product Information
  • Capacity
    Division
    Height
    Ø
    Availability
    Price
  • 10 ml
    0,25 ml
    87 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,70 €
    6,70 €
    1 * 1 ST
       6,70 €
  • 25 ml
    1 ml
    107 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,10 €
    7,10 €
    1 * 1 ST
       7,10 €
  • 50 ml
    2,5 ml
    143 mm
    29 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,30 €
    10,30 €
    1 * 1 ST
       10,30 €
  • 100 ml
    5 ml
    177 mm
    34 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,10 €
    12,10 €
    1 * 1 ST
       12,10 €
  • 250 ml
    10 ml
    263 mm
    45,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,20 €
    28,20 €
    1 * 1 ST
       28,20 €
  • 500 ml
    10 ml
    302 mm
    55,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,30 €
    34,30 €
    1 * 1 ST
       34,30 €
  • 1000 ml
    25 ml
    331 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,90 €
    47,90 €
    1 * 1 ST
       47,90 €
  • 2000 ml
    50 ml
    369 mm
    92 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,80 €
    66,80 €
    1 * 1 ST
       66,80 €