ContactUsLinkComponent
Staining jars acc. to Coplin, WHEATON®
Staining jars acc. to Coplin, WHEATON®
  720-0707
undefined
Staining jars acc. to Coplin, WHEATON®
  720-0707
  • Description:
    Staining jar acc. to Coplin, with screw cap
  • W×D×H:
    26×26×70 mm
  • Pk:
    6
Soda-lime glass.
  • Slides extend above the opening
  • Easy manipulation without using forceps
  • Stable rectangular base
This staining jar has the capacity for up to 10 standard slides of 76×26 mm with white PP screw cap.