Order Entry
Belgium
Orders LinkContactUsLinkComponent
Hydrometer according to Œchsle

Hydrometer according to Œchsle

Hydrometer according to Œchsle, 0 - 130 °, 1 °, 270 mm
Catalog # 614-0515
Supplier:   GERING
Hydrometer according to Œchsle
Catalog # 614-0515
Supplier:  GERING
Hydrometer according to Œchsle
Hydrometer according to Œchsle

Hydrometer according to Œchsle, 0 - 130 °, 1 °, 270 mm

Price
36,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Hydrometer according to Œchsle for grape juices.

Specifications
  • Description:
    Hydrometer according to Œchsle
  • Length:
    270 mm
  • Division:
    1 °
  • Range:
    0 - 130 °