Orders LinkContactUsLinkComponent
Brix saccharimeters
Supplier:   GERING
Brix saccharimeters
Supplier:  GERING
Certificates
Brix saccharimeters
Brix saccharimeters
22 Options Available Below
Product Details & Documents
Brix saccharimeters with a reference temperature of 20 °C.
Product Family Options
Product Information
  • Range
    Division
    Calibration temperature
    Length
    Availability
    Price
  • 0 - 10 °Brix
    0,1
    20 °C
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,20 €
    39,20 €
    1 * 1 ST
       39,20 €
  • 10 - 20 °Brix
    0,1
    20 °C
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,20 €
    39,20 €
    1 * 1 ST
       39,20 €
  • 20 - 30 °Brix
    0,1
    20 °C
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,20 €
    39,20 €
    1 * 1 ST
       39,20 €
  • 50 - 60 °Brix
    0,1
    20 °C
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • 60 - 70 °Brix
    0,1
    20 °C
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • 70 - 80 °Brix
    0,1
    20 °C
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • 80 - 90 °Brix
    0,1
    20 °C
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,60 €
    58,60 €
    1 * 1 ST
       58,60 €
  • 0 - 30 °Brix
    1,0
    20 °C
    230 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,90 €
    32,90 €
    1 * 1 ST
       32,90 €
  • 30 - 60 °Brix
    1,0
    20 °C
    230 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,40 €
    39,40 €
    1 * 1 ST
       39,40 €
  • 60 - 90 °Brix
    1,0
    20 °C
    230 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,40 €
    39,40 €
    1 * 1 ST
       39,40 €
With thermometer
Product Information
  • Range
    Division
    Calibration temperature
    Length
    Availability
    Price
  • 0 - 10 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,50 €
    90,50 €
    1 * 1 ST
       90,50 €
  • 10 - 20 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,50 €
    90,50 €
    1 * 1 ST
       90,50 €
  • 20 - 30 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,50 €
    90,50 €
    1 * 1 ST
       90,50 €
  • 30 - 40 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,50 €
    90,50 €
    1 * 1 ST
       90,50 €
  • 40 - 50 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • 50 - 60 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • 60 - 70 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • 70 - 80 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • 80 - 90 °Brix
    0,1
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • 0 - 30 °Brix
    1,0
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 121,00 €
    121,00 €
    1 * 1 ST
       121,00 €
  • 30 - 60 °Brix
    1,0
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • 60 - 90 °Brix
    1,0
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €