Orders LinkContactUsLinkComponent
Micro-tissue grinder set
Micro-tissue grinder set
Grinder set
Catalog # 432-5063
Supplier:   DWK Life Sciences
Micro-tissue grinder set
Catalog # 432-5063
Supplier:  DWK Life Sciences
Micro-tissue grinder set
Micro-tissue grinder set
Grinder set
Price
989,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
A complete selection of Wheaton Micro tissue grinders conveniently packed in a lightweight protective carrying case.
Includes the following tissue grinders:
- 1 ml Tenbroeck
- 0,5 ml grinder with 13 mm-425 GPI cap
- 1 ml Dounce
- 0,1 and 0,2 ml standard
- 2 ml Potter-Elvehjem
- 1 ml tapered
Specifications
  • Description:
    Grinder set