Orders LinkContactUsLinkComponent
Tissue grinders, conical
Tissue grinders, conical
Supplier:   DWK Life Sciences
Tissue grinders, conical
Supplier:  DWK Life Sciences
Certificates
Tissue grinders, conical
Tissue grinders, conical
12 Options Available Below
Product Details & Documents
Borosilicate glass. Handheld homogeniser with conical surface, which allows initial size reduction, followed by passage through the cylindrical section for the final homogenisation step. Grinding efficiency is improved and less time is required as compared to the Tenbroeck and Potter-Elvehjem designs. Recommended for heart and lung tissues, as well for skin and plant tissues.
  • Clearance between pestle and tube is 0,1 to 0,15 mm
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifications: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Product Family Options
Product Information
  • Capacity
    Ø ext.×length
    Total length
    Availability
    Price
  • 1 ml
    11×49 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • 3 ml
    11×86 mm
    220 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • 5 ml
    13×93 mm
    220 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • 15 ml
    18×114 mm
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 249,00 €
    249,00 €
    1 * 1 ST
       249,00 €
Spare parts
Product Information
  • Capacity
    Ø ext.×length
    Total length
    Availability
    Price
  • 432-5048
    Pestle only, 1 ml
    Pestle only, 1 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 180,00 €
    180,00 €
    1 * 2 ST
       180,00 €
  • 432-5049
    Mortar cylinder only, 1 ml
    Mortar cylinder only, 1 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 232,00 €
    232,00 €
    1 * 2 ST
       232,00 €
  • 432-5050
    Pestle only, 3 ml
    Pestle only, 3 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 180,00 €
    180,00 €
    1 * 2 ST
       180,00 €
  • 432-5051
    Mortar cylinder only, 3 ml
    Mortar cylinder only, 3 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 232,00 €
    232,00 €
    1 * 2 ST
       232,00 €
  • 432-5052
    Pestle only, 5 ml
    Pestle only, 5 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 192,00 €
    192,00 €
    1 * 2 ST
       192,00 €
  • 432-5053
    Mortar cylinder only, 5 ml
    Mortar cylinder only, 5 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 234,00 €
    234,00 €
    1 * 2 ST
       234,00 €
  • 432-5054
    Pestle only, 15 ml
    Pestle only, 15 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 219,00 €
    219,00 €
    1 * 2 ST
       219,00 €
  • 432-5055
    Mortar cylinder only, 15 ml
    Mortar cylinder only, 15 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 265,00 €
    265,00 €
    1 * 2 ST
       265,00 €