Orders LinkContactUsLinkComponent
Micro-grinders
Micro-grinders
Supplier:   DWK Life Sciences
Micro-grinders
Supplier:  DWK Life Sciences
Certificates
Micro-grinders
2 Options Available Below
Product Details & Documents
Borosilicate glass. Designed for delicate manual operation. Similar to all-glass Potter-Elvehjem style, but micro size for extremely precise work.
  • Reservoir and pouring lip incorporated in design
  • Clearance between pestle and tube is 0,1 to 0,15 mm
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifications: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Product Family Options
Product Information
  • Capacity
    Ø ext.×length
    Total length
    Availability
    Price
  • 0,1 ml
    4×65 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 235,00 €
    235,00 €
    1 * 2 ST
       235,00 €
  • 0,2 ml
    7×33 mm
    115 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 185,00 €
    185,00 €
    1 * 2 ST
       185,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Mortar for micro-grinders
    Mortar for micro-grinders
    WHEA357843
    Log in for availability
    1 * 2 ST - 120,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Pestle for micro-grinders
    Pestle for micro-grinders
    WHEA357841
    Log in for availability
    1 * 2 ST - 197,00 €