Orders LinkContactUsLinkComponent
Embedding rings
Embedding rings
Supplier:   Simport Scientific
Embedding rings
Supplier:  Simport Scientific
Certificates
Embedding rings
5 Options Available Below
Product Details & Documents
PS. These embedding rings are suitable for holding and identifying tissue sample blocks and fit well in microtome chuck adapters.
  • The etched writing surface on the ring is marked with an identification number and placed on top of the sample block
  • Additional paraffin is poured into the base mould to cement the ring onto the tissue block
  • The embedding ring securely holds the tissue sample in the microtome chuck adapter for sectioning and then identifies the sample while in storage
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 151,00 €
    151,00 €
    1 * 1.000 ST
       151,00 €
  • Pink
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 151,00 €
    151,00 €
    1 * 1.000 ST
       151,00 €
  • Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 151,00 €
    151,00 €
    1 * 1.000 ST
       151,00 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 151,00 €
    151,00 €
    1 * 1.000 ST
       151,00 €
  • Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 151,00 €
    151,00 €
    1 * 1.000 ST
       151,00 €