Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue capsules

Tissue capsules

Supplier:   Simport Scientific
Tissue capsules
Supplier:  Simport Scientific
Certificates
Tissue capsules

2 Options Available Below

Product Details & Documents

These capsules made of white PP can hold tissue during processing.

  • Frosted area for writing sample ID
  • Lid snaps securely on to the base
Product Family Options

Product Information

  • Colour
    Type
    Availability
    Price
  • White
    38×10 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 452,00 €
    452,00 €
    1 * 1.000 ST
       452,00 €

  • White
    28×5 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 223,00 €
    223,00 €
    1 * 1.000 ST
       223,00 €