Cervical brushes, Cytotak™ Transwab®
Nylon-bristle cytology sampling brushes for cervical smears.
- Plastic shaft is non toxic and safe to use
- Tapered spiral configuration assists access to narrow opening
- Gentle rotation creates a brushing action against skin surface for enhanced recovery of cellular material
Shaft/head: Plastic/nylon
: Available bulk or individually packed.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together