Orders LinkContactUsLinkComponent
Modular Storage Drawers
Modular Storage Drawers
Supplier:   Simport Scientific
Modular Storage Drawers
Supplier:  Simport Scientific
Certificates
Modular Storage Drawers
Modular Storage Drawers
2 Options Available Below
Product Details & Documents
Storage drawers from PS. Permanent storage of up to 165 embedding rings or 250 cassettes per drawer.
  • Interlocking ridges for stacking
  • Identification labels included
Product Family Options
Product Information
  • Availability
    Price
  • 720-0280
    Drawers only, 40,5 × 23 × 5,1 cm
    Drawers only, 40,5 × 23 × 5,1 cm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 173,00 €
    173,00 €
    1 * 6 ST
       173,00 €
  • 720-0281
    Drawers with 1500 embedding cassettes for M480, 40,5 × 23 × 5,1 cm
    Drawers with 1500 embedding cassettes for M480, 40,5 × 23 × 5,1 cm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST