Orders LinkContactUsLinkComponent
Embedding cassettes, M480
Embedding cassettes, M480
Supplier:   Simport Scientific
Embedding cassettes, M480
Supplier:  Simport Scientific
Certificates
Embedding cassettes, M480
12 Options Available Below
Product Details & Documents
Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures.
  • Resistant to all commonly used histological solvents
  • Designed to accept standard metal lids (M481)
  • Slanted writing surface (45° angle) accepts markings easily
Different colours available.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • Purple
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Colourless
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • White
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Pink
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Green
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Blue
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Salmon
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
  • Cream
    Log in for availability
    Log in for availability
    1 * 1.500 Pce
    1 * 1.500 Pce - 213,00 €
    213,00 €
    1 * 1.500 Pce
       213,00 €
  • Grey
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 213,00 €
    213,00 €
    1 * 1.500 ST
       213,00 €
Lids
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Silver
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 138,00 €
    138,00 €
    1 * 25 ST
       138,00 €
Accessories
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • 720-0403
    Silver
    Log in for availability
    1 * 25 ST - 138,00 €