Biopsy processing/embedding cassettes, Micromesh™ M508
Fournisseur: Simport Scientific
Aller à :
À propos de cet article
Acetal polymer.
- No need for biopsy pads
- Cassettes have four square compartments each 13 mm
- Cover and base have about 1676 square openings to maximise fluid exchange
- Integral lid
- Anterior writing area 45° angle suitable for some labelling systems
11 Options disponibles ci-dessous
Product Details & Documents
Acetal polymer.
- No need for biopsy pads
- Cassettes have four square compartments each 13 mm
- Cover and base have about 1676 square openings to maximise fluid exchange
- Integral lid
- Anterior writing area 45° angle suitable for some labelling systems
Product Family Options
Informations sur le produit
- ColourDisponibilitéPrix



