Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Microsette™ M503
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ M503
Supplier:  Simport Scientific
Certificates
Biopsy processing/embedding cassettes, Microsette™ M503
Biopsy processing/embedding cassettes, Microsette™ M503
11 Options Available Below
Product Details & Documents
Acetal polymer.
  • No need for biopsy pads
  • Six compartments, each 7×12 mm, numbered 1 to 6
  • 2000 square openings (each 0,26 mm) to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • Beige
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Purple
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Orange
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Pink
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Salmon
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €
  • Grey
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 524,00 €
    524,00 €
    1 * 1.000 ST
       524,00 €