Orders LinkContactUsLinkComponent
Goggles, Flex Seal™
Goggles, Flex Seal™
Supplier:   HONEYWELL SAFETY
Goggles, Flex Seal™
Supplier:  HONEYWELL SAFETY
Certificates
Goggles, Flex Seal™
Goggles, Flex Seal™
2 Options Available Below
Product Details & Documents
Goggles with a flexible and sturdy frame feature a wraparound lens for a panoramic view.
  • Soft and flexible silicone frame for excellent comfort and ensures a snug, slip-free fit
  • Remains soft and supple even in extremely hot or cold environments
  • Pivoting, removable and adjustable headband
  • Indirect ventilation prevents fogging
  • Neoprene strap (111-0132) or elastic strap (111-3606)
Designed to be compatible with prescription spectacles.
Weight: 110 g.
Certifications: EN 166, EN 169, EN 171, 1FT, C 3, 4, 9.
Product Family Options
Product Information
  • Type
    Lenses
    Colour
    Availability
    Price
  • Anti-UV, anti-scratch, anti-fog
    Clear PC
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,70 €
    32,70 €
    1 * 1 ST
       32,70 €
  • Anti-UV, anti-scratch, anti-fog
    Clear PC
    Blue/grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,00 €
    33,00 €
    1 * 1 ST
       33,00 €