Orders LinkContactUsLinkComponent
Industrial gloves, Perfect Poly® white
Industrial gloves, Perfect Poly® white
Supplier:   HONEYWELL SAFETY
Industrial gloves, Perfect Poly® white
Supplier:  HONEYWELL SAFETY
Certificates
Industrial gloves, Perfect Poly® white
5 Options Available Below
Product Details & Documents
Knitted gloves made of lightweight, white polyamide with PU coating. Suitable for the handling of fine products in a dry or slightly greasy or soiled environment.
  • Polyurethane coating on palm and fingertips
  • Exceptional dexterity
  • High abrasion resistance
  • Non pilling: Suitable for precision work
  • Certified as 'silicone-free' to ensure their use in painting environment
Typical applications include: Automotive industry assembly, logistics (forklift operators), and sorting of parts.
Certifications: EN 388:2016 (3131X); Cat. II
Specifications for Product Family
Specifications
  • Catalog No:
  • DALL2232255/6
  • DALL2232255/7
  • DALL2232255/8
  • DALL2232255/9
  • DALL2232255/10
  • Model:
  • White
  • White
  • White
  • White
  • White
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Finish:
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Gauge:
  • 15
  • 15
  • 15
  • 15
  • 15
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Length
    Availability
    Price
  • 6
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAIRE
    1 * 10 PAIRE
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2232255/6
    Model
    White
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Palm and fingertips coated
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 7
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAIRE
    1 * 10 PAIRE
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2232255/7
    Model
    White
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Palm and fingertips coated
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 8
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAIRE
    1 * 10 PAIRE
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2232255/8
    Model
    White
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Palm and fingertips coated
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 9
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAIRE
    1 * 10 PAIRE
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2232255/9
    Model
    White
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Palm and fingertips coated
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)
  • 10
    White
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAIRE
    1 * 10 PAIRE
    Specifications:
    More
    Specifications
    Cat. No.
    DALL2232255/10
    Model
    White
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Palm and fingertips coated
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (3131X)