Cleanroom Cut-Resistant Glove Liners
Supplier: WELLS LAMONT
Certificates
About this item
Great for use as a cut-resistant cleanroom glove liner.
- Highly reusable
- Made in USA
Certifications: ANSI A4, level C cut resistance. Category III for particulates.
Specifications
- Catalog No:
- 112-1197
- 112-1196
- 112-6365
- 112-1195
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Material:
- Nylon
- Nylon
- Nylon
- Nylon
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile