ContactUsLinkComponent
Examination gloves, neoprene, Micro-Touch® Affinity™
Examination gloves, neoprene, Micro-Touch® Affinity™
Certificates
undefined
Examination gloves, neoprene, Micro-Touch® Affinity™
Specifications
  • Catalog No:
  • 112-1827
  • 112-1826
  • 112-1828
  • 112-1825
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,110 mm
  • 0,110 mm
  • 0,110 mm
  • 0,110 mm
  • Finger thickness:
  • 0,170 mm
  • 0,170 mm
  • 0,170 mm
  • 0,170 mm
  • Palm thickness:
  • 0,130 mm
  • 0,130 mm
  • 0,130 mm
  • 0,130 mm
  • EN norm:
  • EN 455, EN 374
  • EN 455, EN 374
  • EN 455, EN 374
  • EN 455, EN 374