ContactUsLinkComponent
Cleanroom gloves, latex, sterile, Kimtech™ G3
Cleanroom gloves, latex, sterile, Kimtech™ G3
Supplier:  KIMBERLY CLARK
Certificates
undefined
Cleanroom gloves, latex, sterile, Kimtech™ G3
Supplier:  KIMBERLY CLARK
Certificates
Specifications
  • Catalog No:
  • 112-0491
  • 112-4830
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 1/ISO 3
  • 1/ISO 3
  • Cuff style:
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,16 mm
  • 0,16 mm
  • Finger thickness:
  • 0,22 mm
  • 0,22 mm
  • Palm thickness:
  • 0,20 mm
  • 0,20 mm
  • EN norm:
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)
  • EN 374-1 Type C (K), EN 374-5 (VIRUS)