Dropping funnels, cylindrical, without pressure balance, Quickfit®
Supplier: SciLabware
Certificates
About this item
PYREX®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With socket and stem with cone, with GP Rotaflo stopcock.
Frequently Bought Together