Orders LinkContactUsLinkComponent
Cylindrical separating funnels
Cylindrical separating funnels
Supplier:   Lenz Laborglas
Cylindrical separating funnels
Supplier:  Lenz Laborglas
Certificates
Cylindrical separating funnels
Cylindrical separating funnels
10 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With graduation.
Documents
Product Family Options
With solid glass key
Product Information
  • Capacity
    Socket size
    Division
    Availability
    Price
  • 250 ml
    29/32 NS
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,30 €
    75,30 €
    1 * 1 ST
       75,30 €
  • 500 ml
    29/32 NS
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,50 €
    90,50 €
    1 * 1 ST
       90,50 €
  • 50 ml
    29/32 NS
    1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,10 €
    62,10 €
    1 * 1 ST
       62,10 €
  • 100 ml
    29/32 NS
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,10 €
    59,10 €
    1 * 1 ST
       59,10 €
  • 1000 ml
    29/32 NS
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 126,00 €
    126,00 €
    1 * 1 ST
       126,00 €
With PTFE key
Product Information
  • Capacity
    Socket size
    Division
    Availability
    Price
  • 50 ml
    29/32 NS
    1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 74,50 €
    74,50 €
    1 * 1 ST
       74,50 €
  • 100 ml
    29/32 NS
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,40 €
    70,40 €
    1 * 1 ST
       70,40 €
  • 250 ml
    29/32 NS
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,70 €
    94,70 €
    1 * 1 ST
       94,70 €
  • 500 ml
    29/32 NS
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 103,00 €
    103,00 €
    1 * 1 ST
       103,00 €
  • 1000 ml
    29/32 NS
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 137,00 €
    137,00 €
    1 * 1 ST
       137,00 €