Orders LinkContactUsLinkComponent
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®
Supplier:   SciLabware
Büchner funnels, Quickfit®
Supplier:  SciLabware
Certificates
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®
3 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc and integral ground glass cone.
Product Family Options
Product Information
  • Plate Ø
    Capacity
    Porosity
    Cone size
    Availability
    Price
  • 55 mm
    3
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • 65 mm
    3
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €
  • 90 mm
    105 ml
    3
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 132,00 €
    132,00 €
    1 * 1 ST
       132,00 €