Orders LinkContactUsLinkComponent
Filter Funnels, Conical
Filter Funnels, Conical
Supplier:   ROBU GLASFILTER
Filter Funnels, Conical
Supplier:  ROBU GLASFILTER
Certificates
Filter Funnels, Conical
Filter Funnels, Conical
10 Options Available Below
Product Details & Documents
ROBU® filter funnel conical with intergrated VitraPOR® filter is a classic laboratory tool designed for reliable filtration. Made from high-quality borosilicate glass, it ensures chemical resistance and durability across a wide range of applications.
  • Conical design
  • Precise pour control
  • Borosilicate glass
  • Chemical resistance
  • Heat resistance
  • Reusable
  • Suitable for gravity filtration
  • Compatible with standard filter papers
The ROBU funnel with integrated VitraPor filter, conical, is an essential component for laboratory filtration processes. Its tapered design promotes smooth and controlled fluid flow, making it suitable for both qualitative and quantitative filtration. Manufactured from durable borosilicate glass, the funnel offers excellent resistance to thermal shock and aggressive chemicals.
This reusable funnel is compatible with standard filter papers and ideal for a variety of laboratory settings including research, quality control, and educational environments.
Certifications: In compliance with ISO 4796-80 and ASTM E128-99 standards.
Documents
Product Family Options
Product Information
  • Capacity
    Material
    Plate Ø
    Porosity
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 511-3301
    Filter funnel, conical
    Filter funnel, conical
    25 ml
    Borosilicate glass 3.3
    25 mm
    1
    8 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,60 €
    26,60 €
    1 * 1 ST
       26,60 €
  • 511-3302
    Filter funnel, conical
    Filter funnel, conical
    25 ml
    Borosilicate glass 3.3
    25 mm
    2
    8 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,60 €
    26,60 €
    1 * 1 ST
       26,60 €
  • 511-3303
    Filter funnel, conical
    Filter funnel, conical
    25 ml
    Borosilicate glass 3.3
    25 mm
    3
    8 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,60 €
    26,60 €
    1 * 1 ST
       26,60 €
  • 511-3304
    Filter funnel, conical
    Filter funnel, conical
    25 ml
    Borosilicate glass 3.3
    25 mm
    4
    8 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,50 €
    29,50 €
    1 * 1 ST
       29,50 €
  • 511-0378
    Filter funnel, conical
    Filter funnel, conical
    25 ml
    Borosilicate glass 3.3
    25 mm
    5
    8 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,50 €
    44,50 €
    1 * 1 ST
       44,50 €
  • 511-0379
    Filter funnel, conical
    Filter funnel, conical
    170 ml
    Borosilicate glass 3.3
    35 mm
    1
    10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,30 €
    43,30 €
    1 * 1 ST
       43,30 €
  • 511-0380
    Filter funnel, conical
    Filter funnel, conical
    170 ml
    Borosilicate glass 3.3
    35 mm
    2
    10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,30 €
    43,30 €
    1 * 1 ST
       43,30 €
  • 511-0381
    Filter funnel, conical
    Filter funnel, conical
    170 ml
    Borosilicate glass 3.3
    35 mm
    3
    10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,30 €
    43,30 €
    1 * 1 ST
       43,30 €
  • 511-0382
    Filter funnel, conical
    Filter funnel, conical
    170 ml
    Borosilicate glass 3.3
    35 mm
    4
    10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,90 €
    46,90 €
    1 * 1 ST
       46,90 €
  • 511-0383
    Filter funnel, conical
    Filter funnel, conical
    170 ml
    Borosilicate glass 3.3
    35 mm
    5
    10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,30 €
    66,30 €
    1 * 1 ST
       66,30 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.