Orders LinkContactUsLinkComponent
Büchner funnels
Büchner funnels
Certificates
Büchner funnels
Büchner funnels
13 Options Available Below
Product Details & Documents
White glazed porcelain.
For filter paper or membrane filtration discs.
 : DIN 12 905 (511-2502, 511-2501 and 511-2500 not according to DIN).
Product Family Options
Product Information
  • Capacity
    For filter Ø
    Availability
    Price
  • 10 ml
    27 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,70 €
    63,70 €
    1 * 1 ST
       63,70 €
  • 25 ml
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,30 €
    66,30 €
    1 * 1 ST
       66,30 €
  • 50 ml
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,90 €
    70,90 €
    1 * 1 ST
       70,90 €
  • 75 ml
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 78,50 €
    78,50 €
    1 * 1 ST
       78,50 €
  • 135 ml
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 103,00 €
    103,00 €
    1 * 1 ST
       103,00 €
  • 290 ml
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 135,00 €
    135,00 €
    1 * 1 ST
       135,00 €
  • 580 ml
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 168,00 €
    168,00 €
    1 * 1 ST
       168,00 €
  • 795 ml
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 226,00 €
    226,00 €
    1 * 1 ST
       226,00 €
  • 1250 ml
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 311,00 €
    311,00 €
    1 * 1 ST
       311,00 €
  • 1900 ml
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 498,00 €
    498,00 €
    1 * 1 ST
       498,00 €
  • 4300 ml
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 811,00 €
    811,00 €
    1 * 1 ST
       811,00 €
  • 5800 ml
    270 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 510,00 €
    1 510,00 €
    1 * 1 ST
       1 510,00 €
  • 10600 ml
    320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 990,00 €
    1 990,00 €
    1 * 1 ST
       1 990,00 €